missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(cyclopropylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15490196
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490196 1 g 1g
15400206 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490196 Supplier Thermo Scientific Alfa Aesar Supplier No. H52910.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850589-44-5
Molecular Formula C10H11BClNO3
Molecular Weight (g/mol) 239.462
MDL Number MFCD07363770
InChI Key TUPWHDSMIIRKLY-UHFFFAOYSA-N
Synonym 3-chloro-4-cyclopropylcarbamoyl phenylboronic acid, 3-chloro-4-cyclopropylcarbamoyl phenyl boronic acid, 3-chloro-4-cyclopropylcarbamoyl benzeneboronic acid, n-cyclopropyl 2-chloro-4-boronobenzamide, 3-chloro-4-cyclopropylcarbamoyl phenyl boronicacid, 3-chloro-4-cyclopropylcarbamoyl-phenyl boronic acid, 3-chloro-4-cyclopropylamino carbonyl phenyl boronic acid, boronic acid, b-3-chloro-4-cyclopropylamino carbonyl phenyl, boronic acid,b-3-chloro-4-cyclopropylamino carbonyl phenyl
PubChem CID 44119649
IUPAC Name [3-chloro-4-(cyclopropylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC(=C(C=C1)C(=O)NC2CC2)Cl)(O)O

Specifications

Melting Point 216°C to 220°C
Quantity 1 g
Formula Weight 239.46
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(cyclopropylcarbamoyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.