Learn More
3-Chloro-1-(4-methoxyphenyl)propan-1-one, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-1-(4-methoxyphenyl)propan-1-one |
| CAS | 35999-20-3 |
| Molecular Formula | C10H11O2Cl |
| Quantity | 5 g |
| Synonym | 3-Chloro-1-(4-methoxyphenyl)-1-propanone, 2-Chloroethyl 4-Methoxyphenyl Ketone, 3-Chloro-1-(4'-methoxyphenyl)propan-1-one, 3-Chloro-4'-methoxypropiophenone, NSC 31620, p-(3-Chloropropan-1-one)anisole, p-Methoxy-β-chloropropiophenone, β-Chloro-p-methoxypropiophenone |
| InChI Formula | InChI=1S/C10H11ClO2/c1-13-9-4-2-8(3-5-9)10(12)6-7-11/h2-5H,6-7H2,1H3 |
| SMILES | COc1ccc(cc1)C(=O)CCCl |
| IUPAC Name | 3-chloro-1-(4-methoxyphenyl)propan-1-one |
| Molecular Weight (g/mol) | 198.65 |