Learn More
3-Carboxyphenyl isothiocyanate, 97%
Product Code. 11327706
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327706 5 g 5g
11317706 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11327706 Supplier Thermo Scientific Alfa Aesar Supplier No. L11322.06
 Restricted Item
In Stock
Add to basket
CAS: 2131-63-7 | C8H5NO2S | 179.193 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 2131-63-7
Molecular Formula C8H5NO2S
Molecular Weight (g/mol) 179.193
MDL Number MFCD00041367
InChI Key PJRBPKOOGLKPFB-UHFFFAOYSA-N
Synonym 3-carboxyphenyl isothiocyanate, 3-carboxyphenylisothiocyanate, benzoic acid, 3-isothiocyanato, benzoic acid,3-isothiocyanato, acmc-20an7x, m-carboxyphenyl isothiocyanate, 3-isothiocyanato-benzoic acid, #
PubChem CID 75046
IUPAC Name 3-isothiocyanatobenzoic acid
SMILES C1=CC(=CC(=C1)N=C=S)C(=O)O
Spezifikation
Melting Point ∽165°C (decomposition)
Odor Irritating
Quantity 5 g
UN Number UN2923
Beilstein 2209003
Sensitivity Moisture sensitive
Formula Weight 179.2
Percent Purity 97%
Chemical Name or Material 3-Carboxyphenyl isothiocyanate