Learn More
3-Carboxy-PROXYL, free radical, 97+%
Description
3-Carboxy-PROXYL is used in the synthesis and reactions of stable nitroxyl radicals. It is also used to study molecular migration and long-term stability of seeds and pollen and used in the preparation of a tetraphenylporphyrin bearing a nitroxyl radical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Carboxy-PROXYL is used in the synthesis and reactions of stable nitroxyl radicals. It is also used to study molecular migration and long-term stability of seeds and pollen and used in the preparation of a tetraphenylporphyrin bearing a nitroxyl radical.
Solubility
Soluble in water (partly miscible), methanol (50 mg/ml), acetone, dichloromethane and chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.
Specifications
Specifications
| Melting Point | ∽200°C (decomposition) |
| Quantity | 50 mg |
| Beilstein | 4136411 |
| Solubility Information | Soluble in water (partly miscible),methanol (50mg/ml),acetone,dichloromethane and chloroform. |
| Formula Weight | 186.23 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-Carboxy-PROXYL, free radical |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.