accessibility menu, dialog, popup

UserName

CAS RN 2154-68-9

CH3H3C(R)OOCH3H3CNHO

CAS RN 2154-68-9

IUPAC Name: (3R)-1-hydroxy-2,2,5,5-tetramethylpyrrolidine-3-carboxylate
Synonyms: (3R)-1-hydroxy-2,2,5,5-tetramethylpyrrolidine-3-carboxylate
Molecular Weight (g/mol): 186.23
Molecular Formula: C9H16NO3
InChi Key: CLKPVQZFNYXFCY-LURJTMIESA-M
SMILES: CC1(C)C[C@@H](C([O-])=O)C(C)(C)N1O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results