Learn More
(3-Bromopropyl)triphenylphosphonium bromide, 98%, Thermo Scientific™
Product Code. 11439973
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439973 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11439973 Supplier Thermo Scientific Alfa Aesar Supplier No. A13041.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3607-17-8
Molecular Formula C21H21Br2P
Molecular Weight (g/mol) 464.181
MDL Number MFCD00011866
InChI Key ZAHUZZUGJRPGKW-UHFFFAOYSA-M
Synonym 3-bromopropyl triphenylphosphonium bromide, 3-bromopropyltriphenylphosphonium bromide, bromo 3-bromopropyl triphenylphosphorus, phosphonium, 3-bromopropyl triphenyl-, bromide, 3-bromopropyl triphenylphosphanium bromide, 3-bromopropyl triphenylphosphonium, acmc-209ikr, bromo 3-bromopropyl triphenylphosphorane, 3-brompropyl triphenylphosphonium bromide
PubChem CID 2723862
IUPAC Name 3-bromopropyl(triphenyl)phosphanium;bromide
SMILES C1=CC=C(C=C1)[P+](CCCBr)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
Specifications
Melting Point 225°C to 229°C
Quantity 100 g
Beilstein 6422974
Formula Weight 464.19
Percent Purity 98%
Chemical Name or Material (3-Bromopropyl)triphenylphosphonium bromide