missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzaldehyde diethyl acetal, 97%, Thermo Scientific™

Product Code. 11338296
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338296 5 g 5g
11348296 25 g 25g
11358296 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338296 Supplier Thermo Scientific Alfa Aesar Supplier No. L15514.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75148-49-1
Molecular Formula C11H15BrO2
Molecular Weight (g/mol) 259.14
MDL Number MFCD01075697
InChI Key KDHRJLQXMOBXRV-UHFFFAOYSA-N
Synonym 3-bromobenzaldehyde diethyl acetal, 1-bromo-3-diethoxymethyl benzene, 3-bromophenyl diethoxymethane, benzene, 1-bromo-3-diethoxymethyl, 3-bromo-alpha,alpha-diethoxytoluene, 3-bromobenzaldehydediethylacetal, pubchem5371, acmc-209ox6, 1-diethoxymethyl-3-bromobenzene, 1-bromo-3-diethoxymethyl-benzene
PubChem CID 2733543
IUPAC Name 1-bromo-3-(diethoxymethyl)benzene
SMILES CCOC(OCC)C1=CC=CC(Br)=C1

Specifications

Density 1.271
Boiling Point 88°C to 91°C (0.8 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.514
Quantity 5 g
Beilstein 6392356
Formula Weight 259.14
Percent Purity 97%
Chemical Name or Material 3-Bromobenzaldehyde diethyl acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.