Learn More
3-Bromo-N,N-dimethylaniline, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-N,N-dimethylaniline |
| CAS | 16518-62-0 |
| Molecular Formula | C8 H10 Br N |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C8H10BrN/c1-10(2)8-5-3-4-7(9)6-8/h3-6H,1-2H3 |
| SMILES | CN(C)c1cccc(Br)c1 |
| IUPAC Name | 3-bromo-N,N-dimethylaniline |
| Molecular Weight (g/mol) | 200.08 |
| Formula Weight | 198.9997 |