Learn More
3-Bromo-4-isopropylbenzoic acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-4-isopropylbenzoic acid |
| CAS | 99070-17-4 |
| Molecular Formula | C10H11BrO2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C10H11BrO2/c1-6(2)8-4-3-7(10(12)13)5-9(8)11/h3-6H,1-2H3,(H,12,13) |
| SMILES | CC(C)c1ccc(cc1Br)C(=O)O |
| IUPAC Name | 3-bromo-4-propan-2-ylbenzoic acid |
| Molecular Weight (g/mol) | 243.1 |
| Formula Weight | 241.9942 |