missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-nitrobenzaldehyde, TRC

Product Code. 30348734
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348734 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348734 Supplier Toronto Research Chemicals Supplier No. TRCB68726525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Bromo-2-nitrobenzaldehyde
CAS 882772-99-8
Molecular Formula C7H4BrNO3
Quantity 25 g
InChI Formula InChI=1S/C7H4BrNO3/c8-6-3-1-2-5(4-10)7(6)9(11)12/h1-4H
SMILES [O-][N+](=O)c1c(Br)cccc1C=O
IUPAC Name 3-bromo-2-nitrobenzaldehyde
Molecular Weight (g/mol) 230.02
Formula Weight 228.9375
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.