missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-methylpyrrolidine, 97%, Thermo Scientific™

Product Code. p-7056988
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492709 1 g 1g
15402719 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15492709 Supplier Thermo Scientific Alfa Aesar Supplier No. H66250.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10603-45-9
Molecular Formula C5H10BrN
Molecular Weight (g/mol) 164.046
MDL Number MFCD12755877
InChI Key ZHUYTHOKJZSZQJ-UHFFFAOYSA-N
Synonym 3-bromo-1-methyl-pyrrolidine, pyrrolidine, 3-bromo-1-methyl, 3-bromo-1-methylpyrrolidine
PubChem CID 13726279
IUPAC Name 3-bromo-1-methylpyrrolidine
SMILES CN1CCC(C1)Br

Specifications

Color Yellow to Brown
Density 1.361 g/mL
Flash Point 54°C (129°F)
Refractive Index 1.491
Quantity 1 g
UN Number UN2733
Sensitivity Moisture sensitive
Formula Weight 164.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromo-1-methylpyrrolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.