missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)azetidine hydrochloride, 97%, Thermo Scientific™

Product Code. p-7040076
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426088 250 mg 250mg
15436088 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15426088 Supplier Thermo Scientific Alfa Aesar Supplier No. H59695.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 217806-26-3
Molecular Formula C8H17ClN2O2
Molecular Weight (g/mol) 208.686
MDL Number MFCD09839423
InChI Key VLOLOLUAVFGUTD-UHFFFAOYSA-N
Synonym 3-boc-aminoazetidine hydrochloride, tert-butyl azetidin-3-ylcarbamate hydrochloride, 3-n-boc-aminoazetidine hcl, tert-butyl azetidin-3-yl carbamate hydrochloride, tert-butyl n-azetidin-3-yl carbamate hydrochloride, 3-boc-amino azetidine hydrochloride, 3-n-boc-aminoazetidine hydrochloride, 3-boc-amino-azetidine hcl, tert-butyl 3-azetidinylcarbamate hydrochloride, 3-n-boc-amino-azetidine-hcl
PubChem CID 22101213
IUPAC Name tert-butyl N-(azetidin-3-yl)carbamate;hydrochloride
SMILES CC(C)(C)OC(=O)NC1CNC1.Cl

Specifications

Boiling Point 303°C
Quantity 250 mg
Flash Point 137°C (279°F)
Formula Weight 208.69
Percent Purity 97%
Chemical Name or Material 3-(Boc-amino)azetidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.