missing translation for 'onlineSavingsMsg'
Learn More

3-Biphenylboronic Acid, TRC

Product Code. 30342119
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30342119 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342119 Supplier Toronto Research Chemicals Supplier No. TRCB53522810G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Biphenylboronic Acid
CAS 5122-95-2
Molecular Formula C12H11BO2
Quantity 10 g
Synonym [1,1'-Biphenyl]-3-ylboronic Acid, B-[1,1'-Biphenyl]-3-ylboronic Acid, (3-Phenylphenyl)boronic Acid, (Biphenyl-3-yl)boronic Acid, 1,1'-Biphenyl-3-boronic Acid, 1,1'-Biphenyl-3-ylboronic Acid, 3-(Dihydroxyboryl)-1,1'-biphenyl, 3-Phenylbenzeneboronic Acid
InChI Formula InChI=1S/C12H11BO2/c14-13(15)12-8-4-7-11(9-12)10-5-2-1-3-6-10/h1-9,14-15H
SMILES OB(O)c1cccc(c1)c2ccccc2
IUPAC Name (3-phenylphenyl)boronic acid
Molecular Weight (g/mol) 198.03
Formula Weight 198.0852
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.