Learn More
3-Benzylaniline, 98%, Thermo Scientific™
Product Code. 11339885
Change view
Click to view available options
Quantity:
1 g
5 g
Packungsgröße:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
11339885 1 g 1g
11349885 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11339885 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. L19872.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 61424-26-8
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.254
MDL Number MFCD01310782
InChI Key GZYMMTQDXHJALZ-UHFFFAOYSA-N
PubChem CID 308869
IUPAC Name 3-benzylaniline
SMILES C1=CC=C(C=C1)CC2=CC(=CC=C2)N
Spezifikation
Melting Point 48°C to 52°C
Boiling Point 126°C (2 mmHg)
Quantity 1 g
Beilstein 2803261
Synonym 3-benzyl aniline, benzenamine, 3-phenylmethyl, benzenamine,3-phenylmethyl, 3-benzylphenylamine, 3-phenylmethyl aniline, 3-benzylaniline, 3-phenylmethyl benzenamine, acmc-1b59e
Formula Weight 183.25
Percent Purity 98%
Chemical Name or Material 3-Benzylaniline