Learn More
(3-Azidopropyl)triphenylphosphonium Bromide, TRC
Product Code. 30011803
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011803 250 mg 250mg
30009097 500 mg 500mg
30010747 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011803 Supplier Toronto Research Chemicals Supplier No. TRCA922790250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 127611-39-6
Molecular Formula C21 H21 N3 P . Br
Molecular Weight (g/mol) 426.29
Synonym (3-Azidopropyl)triphenyl-phosphonium Bromide
IUPAC Name 3-azidopropyl(triphenyl)phosphonium;bromide
SMILES [Br-].[N-]=[N+]=NCCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C21H21N3P.BrH/c22-24-23-17-10-18-25(19-11-4-1-5-12-19,20-13-6-2-7-14-20)21-15-8-3-9-16-21;/h1-9,11-16H,10,17-18H2;1H/q+1;/p-1
Formula Weight 425.0656
Chemical Name or Material (3-Azidopropyl)triphenylphosphonium Bromide