Learn More
3-Azido-3-deoxy-4-hydroxy-methyl-1,2-O-isopropylidene-Alpha-D-ribofuranose, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Azido-3-deoxy-4-hydroxy-methyl-1,2-O-isopropylidene-a-D-ri |
| CAS | 247025-10-1 |
| Molecular Formula | C9 H15 N3 O5 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C9H15N3O5/c1-8(2)15-5-6(11-12-10)9(3-13,4-14)17-7(5)16-8/h5-7,13-14H,3-4H2,1-2H3/t5-,6+,7+/m1/s1 |
| SMILES | CC1(C)O[C@H]2OC(CO)(CO)[C@@H](N=[N+]=[N-])[C@H]2O1 |
| IUPAC Name | [(3aR,6S,6aR)-6-azido-5-(hydroxymethyl)-2,2-dimethyl-6,6a-dihydro-3aH-furo[2,3-d][1,3]dioxol-5-yl]methanol |
| Molecular Weight (g/mol) | 245.23 |
| Formula Weight | 245.1012 |