Learn More
3-Aminopiperidine-2,6-dione Hydrochloride, TRC
Product Code. 30132571
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132571 25 g 25g
30009931 1 g 1g
30009455 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132571 Supplier Toronto Research Chemicals Supplier No. TRCA63298025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24666-56-6
Molecular Formula C5H8N2O2.ClH
Molecular Weight (g/mol) 164.59
Synonym 3-Amino-2,6-piperidinedione Monohydrochloride, 2-Amino-glutarimide Hydrochloride, 3-Amino-2,6-dioxopiperidine Hydrochloride, 3-Amino-2,6-piperidinedione Hydrochloride, 3-Aminopiperidine-2,6-dione Monohydrochloride; DL-Glutamic Acid Imide Hydrochloride, Glutamimide Hydrochloride
IUPAC Name (3S)-3-aminopiperidine-2,6-dione;hydrochloride
SMILES Cl.N[C@H]1CCC(=O)NC1=O
Specifications
Analyte or Component Names 3-Aminopiperidine-2,6-dione Hydrochloride
Shipping Condition Room Temperature
InChI Formula InChI=1S/C5H8N2O2.ClH/c6-3-1-2-4(8)7-5(3)9;/h3H,1-2,6H2,(H,7,8,9);1H/t3-;/m0./s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 25 g
Formula Weight 164.0353
Percent Purity >95%
Chemical Name or Material 3-Aminopiperidine-2,6-dione Hydrochloride
Grade HPLC