missing translation for 'onlineSavingsMsg'
Learn More

3-(Aminomethyl)-5-chloroaniline (~90%), TRC

Product Code. 30338018
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338018 50 mg 50mg
30340877 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30338018 Supplier Toronto Research Chemicals Supplier No. TRCA63649050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Aminomethyl)-5-chloroaniline
CAS 683740-35-4
Molecular Formula C7 H9 Cl N2
Quantity 50 mg
Synonym 3-Amino-5-chlorobenzenemethanamine, 3-Amino-5-chlorobenzylamine
InChI Formula InChI=1S/C7H9ClN2/c8-6-1-5(4-9)2-7(10)3-6/h1-3H,4,9-10H2
SMILES NCc1cc(N)cc(Cl)c1
IUPAC Name 3-(aminomethyl)-5-chloroaniline
Molecular Weight (g/mol) 156.613
Formula Weight 156.045
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.