missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4,4,4-trifluorobutyric acid hydrochloride, 95%, Thermo Scientific™

Product Code. 15470847
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470847 1 g 1g
15480847 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470847 Supplier Thermo Scientific Alfa Aesar Supplier No. L18336.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91291-66-6
Molecular Formula C4H7ClF3NO2
Molecular Weight (g/mol) 193.55
MDL Number MFCD00181951
InChI Key MRMARVCQZOYQPI-UHFFFAOYNA-N
Synonym 3-amino-4,4,4-trifluorobutanoic acid hydrochloride, 3-amino-4,4,4-trifluorobutyric acid hydrochloride
PubChem CID 14442472
IUPAC Name 3-amino-4,4,4-trifluorobutanoic acid;hydrochloride
SMILES Cl.NC(CC(O)=O)C(F)(F)F

Specifications

Melting Point 197°C to 199°C
Quantity 1 g
Formula Weight 193.55
Percent Purity 95%
Chemical Name or Material 3-Amino-4,4,4-trifluorobutyric acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.