missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, 99%

Product Code. p-58899 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2 g
Packaging:
Glass bottle
Unit Size:
2g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10229550 500 mg Glass bottle 500mg
10156803 2 g Glass bottle 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10229550 Supplier Thermo Scientific Acros Supplier No. 401105000
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34272-83-8 | C8H17N2O | 157.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34272-83-8
Molecular Formula C8H17N2O
Molecular Weight (g/mol) 157.24
MDL Number MFCD00046088
InChI Key USGBLEMQARVRDC-UHFFFAOYSA-N
Synonym 3s-1-hydroxy-2,2,5,5-tetramethylpyrrolidin-3-aminium
PubChem CID 558590
IUPAC Name 1-hydroxy-2,2,5,5-tetramethylpyrrolidin-3-amine
SMILES CC1(CC(C(N1O)(C)C)N)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 45.0°C
Color Orange
Boiling Point 75.0°C to 80.0°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 500 mg
Formula Weight 157.24
Percent Purity 99%
Physical Form Hygroscopic Crystals
Chemical Name or Material 3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.