Learn More
3-Amino-1-propanol, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | 3-Amino-1-propanol |
| CAS | 156-87-6 |
| Boiling Range | 187°C to 190°C |
| Density | 0.99 g/cm³ at 20°C |
| Flash Point | 100°C (Pensky-Martens closed cup) |
| Melting Range | 11.5°C to 12.4°C |
| Quantity | 25 kg |
| Synonym | 3-Amino-1-propanol, 3-Amino-1-propanol |
| Molecular Weight (g/mol) | 75.11 |
| Grade | Not Available |