missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1,2,4-triazole, 96.1%, MP Biomedicals™

Product Code. 11407770 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 11407770

Brand: MP Biomedicals 0210064710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 61-82-5
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.08
MDL Number MFCD00005230,MFCD00053362
InChI Key KLSJWNVTNUYHDU-UHFFFAOYSA-N
Synonym 3-amino-1,2,4-triazole, amitrole, aminotriazole, 4h-1,2,4-triazol-3-amine, 1h-1,2,4-triazol-3-amine, amitrol, amerol, amizol, emisol, vorox
PubChem CID 1639
ChEBI CHEBI:40036
IUPAC Name 1H-1,2,4-triazol-5-amine
SMILES NC1=NC=NN1

Specifications

Color White
Density ≥0.6 to 0.85g/cm3 at 20°C
Assay Percent Range 0.961
Quantity 10 g
Formula Weight 84.08
Physical Form Powder
Chemical Name or Material 3-Amino-1, 2, 4-triazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.