Learn More
3-Amino-1,1,1-trifluoropropan-2-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Amino-1,1,1-trifluoropropan-2-ol |
| CAS | 431-38-9 |
| Molecular Formula | C3H6F3NO |
| Quantity | 500 mg |
| Synonym | 3-Amino-1,1,1-trifluoro-2-propanol, 1-Amino-3,3,3-trifluoropropan-2-ol, 2-Amino-1-(trifluoromethyl)ethanol, 2-Hydroxy-2-(trifluoromethyl)ethylamine, 2-Hydroxy-3,3,3-trifluoropropylamine |
| InChI Formula | InChI=1S/C3H6F3NO/c4-3(5,6)2(8)1-7/h2,8H,1,7H2 |
| SMILES | NCC(O)C(F)(F)F |
| IUPAC Name | 3-amino-1,1,1-trifluoropropan-2-ol |
| Molecular Weight (g/mol) | 129.08 |
| Formula Weight | 129.0401 |