missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylbenzophenone, TRC

Product Code. 30136406
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136406 100 mg 100mg
30004624 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136406 Supplier Toronto Research Chemicals Supplier No. TRCA170975100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66067-44-5
Molecular Formula C15 H12 O2
Molecular Weight (g/mol) 224.25
Synonym 1-(3-Benzoylphenyl)ethanone, Ketoprofen Imp. A (EP), 3-Acetylbenzophenone
IUPAC Name 1-(3-benzoylphenyl)ethanone
SMILES CC(=O)c1cccc(c1)C(=O)c2ccccc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H12O2/c1-11(16)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-10H,1H3
Formula Weight 224.0837
Percent Purity >95%
Chemical Name or Material 3-Acetylbenzophenone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.