Learn More
3-Acetoxy-2-butanone, 98%
Product Code. 11397046
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397046 5 g 5g
11307056 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11397046 Supplier Thermo Scientific Alfa Aesar Supplier No. L06927.06
Request Bulk or Custom Format
CAS: 4906-24-5 | C6H10O3 | 130.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4906-24-5
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00051782
InChI Key ZKPTYCJWRHHBOW-UHFFFAOYNA-N
Synonym 3-acetoxy-2-butanone, acetoin acetate, 2-acetoxy-3-butanone, 2-butanone, 3-acetyloxy, 3-acetyloxy-2-butanone, 2-butanon-3-ol, acetate, acetic acid 1-methyl-2-oxopropyl ester, 2-butanon-3-yl acetate, 3-hydroxy-2-butanone acetate, acetyl methyl carbinyl acetate
PubChem CID 521246
IUPAC Name 3-oxobutan-2-yl acetate
SMILES CC(OC(C)=O)C(C)=O
Specifications
Density 1.025
Boiling Point 154°C to 155°C
Flash Point 66°C (150°F)
Refractive Index 1.4135
Quantity 5 g
Beilstein 1756882
Formula Weight 130.14
Percent Purity 98%
Chemical Name or Material 3-Acetoxy-2-butanone