Learn More
3,7,4'-Trihydroxyflavone, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Deoxykaempferol |
| CAS | 2034-65-3 |
| Molecular Formula | C15 H10 O5 |
| Quantity | 100 mg |
| Synonym | 4',7-Dihydroxyflavanol, 3,4',7-Trihydroxyflavone, Resokaempferol |
| InChI Formula | InChI=1S/C15H10O5/c16-9-3-1-8(2-4-9)15-14(19)13(18)11-6-5-10(17)7-12(11)20-15/h1-7,16-17,19H |
| SMILES | OC1=C(Oc2cc(O)ccc2C1=O)c3ccc(O)cc3 |
| IUPAC Name | 3,7-dihydroxy-2-(4-hydroxyphenyl)chromen-4-one |
| Molecular Weight (g/mol) | 270.24 |
| Formula Weight | 270.0528 |