missing translation for 'onlineSavingsMsg'
Learn More

(3,5-Dimethoxyphenyl)acetonitrile, TRC

Product Code. 30378267
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378267 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378267 Supplier Toronto Research Chemicals Supplier No. TRCD460585500MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3,5-Dimethoxyphenyl)acetonitrile
CAS 13388-75-5
Molecular Formula C10H11NO2
Quantity 500 mg
Synonym 3,5-Dimethoxy-benzeneacetonitrile, 3,5-Dimethoxybenzyl Cyanide, NSC 245126
InChI Formula InChI=1S/C10H11NO2/c1-12-9-5-8(3-4-11)6-10(7-9)13-2/h5-7H,3H2,1-2H3
SMILES COc1cc(CC#N)cc(OC)c1
IUPAC Name 2-(3,5-dimethoxyphenyl)acetonitrile
Molecular Weight (g/mol) 177.2
Formula Weight 177.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.