missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodosalicylic acid lithium salt hydrate, 95%, Thermo Scientific™

Product Code. 11462448
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
11462448 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 11462448

Brand: Thermo Scientific Alfa Aesar A15784.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 653-14-5
Molecular Formula C7H3I2LiO3
Molecular Weight (g/mol) 395.85
MDL Number MFCD00156142,MFCD00002443
InChI Key HLBRJWWTLIAOTE-UHFFFAOYSA-M
Synonym lithium 3,5-diiodosalicylate, lithium 2-hydroxy-3,5-diiodobenzoate, benzoic acid, 2-hydroxy-3,5-diiodo-, monolithium salt, 3,5-diiodosalicylic acid lithium salt, 2-hydroxy-3,5-diiodobenzoic acid lithium salt, 3,5-diiodo-2-hydroxybenzoic acid lithium salt, benzoic acid, 2-hydroxy-3,5-diiodo-, lithium salt 1:1, 3,5-diiodosalicylic acid lithium salt hydrate, acmc-1ark0, 2-hydroxy-3,5-diiodobenzoic acid, lithium salt
PubChem CID 2735070
SMILES [Li+].OC1=C(I)C=C(I)C=C1C([O-])=O

Specifications

Quantity 25 g
IUPAC Name lithium(1+) 2-hydroxy-3,5-diiodobenzoate
Formula Weight 413.86 (395.85 Anhydrous)
Percent Purity 95%
Chemical Name or Material 3,5-Diiodosalicylic acid lithium salt hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.