accessibility menu, dialog, popup

UserName

LiOHIIOO

CAS RN 653-14-5

IUPAC Name: lithium(1+) 2-hydroxy-3,5-diiodobenzoate
Synonyms: lithium(1+) 2-hydroxy-3,5-diiodobenzoate
Molecular Weight (g/mol): 395.85
Molecular Formula: C7H3I2LiO3
InChi Key: HLBRJWWTLIAOTE-UHFFFAOYSA-M
SMILES: [Li+].OC1=C(I)C=C(I)C=C1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results