Learn More
3,5-Diiodosalicylaldehyde, 98+%
Product Code. 11337595
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337595 10 g 10g
11347595 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11337595 Supplier Thermo Scientific Alfa Aesar Supplier No. L05783.09
Request Bulk or Custom Format
CAS: 2631-77-8 | C7H4I2O2 | 373.916 g/mol
3,5-Diiodosalicylaldehyde is used to synthesize new Schiff bases, (2,4-diiodo-6-[(2-morpholin-4-yl-ethylimino)-methyl]-phenol and 2,4-diiodo-6-[(3-morpholin-4-yl-propylimino)-methyl]-phenol) and the new tridentate ligand, [(2-hydroxy-3,5-diiodo-benzylidene)-amino]-acetic acid (HDBA), 3-bromo-N?-(2-hydroxy-3,5-diiodobenzylidene)benzohydrazide monohydrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Diiodosalicylaldehyde is used to synthesize new Schiff bases, (2,4-diiodo-6-[(2-morpholin-4-yl-ethylimino)-methyl]-phenol and 2,4-diiodo-6-[(3-morpholin-4-yl-propylimino)-methyl]-phenol) and the new tridentate ligand, [(2-hydroxy-3,5-diiodo-benzylidene)-amino]-acetic acid (HDBA), 3-bromo-N′-(2-hydroxy-3,5-diiodobenzylidene)benzohydrazide monohydrate.
Solubility
Insoluble in water.
Notes
Air & Light Sensitive. Store in the dark. Incompatible with light and oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2631-77-8
Molecular Formula C7H4I2O2
Molecular Weight (g/mol) 373.916
MDL Number MFCD00003321
InChI Key MYWSBJKVOUZCIA-UHFFFAOYSA-N
PubChem CID 75829
IUPAC Name 2-hydroxy-3,5-diiodobenzaldehyde
SMILES C1=C(C=C(C(=C1I)O)C=O)I
Specifications
Melting Point 106°C to 110°C
Quantity 10 g
Beilstein 1102552
Sensitivity Air and light sensitive
Synonym 3,5-diiodosalicylaldehyde, benzaldehyde, 2-hydroxy-3,5-diiodo, 3,5-diiodo-2-hydroxybenzaldehyde, 2-hydroxy-3,5-diiodo-benzaldehyde, acmc-20ancp, 3,5-diodosolicylaldehyde, 2,4-diiodo-6-formylphenol, #, benzaldehyde,2-hydroxy-3,5-diiodo
Solubility Information Insoluble in water.
Formula Weight 373.92
Percent Purity ≥98%
Chemical Name or Material 3,5-Diiodosalicylaldehyde