missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodo-L-tyrosine, TRC

Product Code. 30009422
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009422 1 g 1g
30007817 10 g 10g
30008615 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009422 Supplier Toronto Research Chemicals Supplier No. TRCD4660551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 300-39-0
Molecular Formula C9 H9 I2 N O3
Molecular Weight (g/mol) 432.98
Synonym L-Tyrosine, 3,5-diiodo-, Tyrosine, 3,5-diiodo-, L- (8CI), 3,5-Diiodo-L-tyrosine, 3',5'-Diiodo-L-tyrosine, Bijodtyrosin, DIT, DIT (amino acid), L-3,5-Diiodotyrosine, NSC 4143, Liothyronine EP impurity B
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid
SMILES N[C@@H](Cc1cc(I)c(O)c(I)c1)C(=O)O

Specifications

InChI Formula InChI=1S/C9H9I2NO3/c10-5-1-4(2-6(11)8(5)13)3-7(12)9(14)15/h1-2,7,13H,3,12H2,(H,14,15)/t7-/m0/s1
Quantity 1 g
Formula Weight 432.8672
Shipping Condition Room Temperature
Chemical Name or Material Di-Iodo-L-tyrosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.