Learn More
3,5-Difluorothiophenol, TRC
Product Code. 30387087
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387087 50 mg
30378946 100 mg
30370775 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30387087 Supplier Toronto Research Chemicals Supplier No. TRCD45208050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 99389-26-1
Molecular Formula C6H4F2S
Molecular Weight (g/mol) 146.15
IUPAC Name 3,5-difluorobenzenethiol
SMILES SC1=CC(F)=CC(F)=C1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C6H4F2S/c7-4-1-5(8)3-6(9)2-4/h1-3,9H
Formula Weight 146
Chemical Name or Material 3,5-Difluorothiophenol