Learn More
3,5-Dichloropyridine, 98%
Product Code. 11382126
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382126 100 g 100g
11372126 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11382126 Supplier Thermo Scientific Alfa Aesar Supplier No. L03380.22
Request Bulk or Custom Format
CAS: 2457-47-8 | C5H3Cl2N | 147.99 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2457-47-8
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006376
InChI Key WPGHPGAUFIJVJF-UHFFFAOYSA-N
Synonym pyridine, 3,5-dichloro, ccris 1719, 3.5-dichloropyridine, 3,5-dichloro pyridine, 3,5-dichloro-pyridine, pubchem13506, acmc-1cqt0, wln: t6nj cg eg, 5-20-05-00417 beilstein handbook reference, ksc493c0d
PubChem CID 17153
IUPAC Name 3,5-dichloropyridine
SMILES ClC1=CC(Cl)=CN=C1
Specifications
Melting Point 63°C to 67°C
Density 1.39
Boiling Point 178°C
Flash Point 74°C (165°F)
Odor Unpleasant
Quantity 100 g
UN Number UN2811
Beilstein 1973
Formula Weight 147.99
Percent Purity 98%
Chemical Name or Material 3,5-Dichloropyridine