Learn More
3,5-Dichlorophenoxyacetonitrile, 97%, Thermo Scientific™
Product Code. 11398455
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398455 5 g 5g
11388455 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11398455 Supplier Thermo Scientific Alfa Aesar Supplier No. L10490.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103140-12-1
Molecular Formula C8H5Cl2NO
Molecular Weight (g/mol) 202.03
MDL Number MFCD00052707
InChI Key PNROREDTZJCOHF-UHFFFAOYSA-N
Synonym 2-3,5-dichlorophenoxy acetonitrile, 3,5-dichlorophenoxyacetonitrile, 2-3,5-dichlorophenoxy ethanenitrile, maybridge1_008353, acmc-1bu1u, 3,5-dichlorophenoxy acetonitrile, acetonitrile,2-3,5-dichlorophenoxy
PubChem CID 517810
IUPAC Name 2-(3,5-dichlorophenoxy)acetonitrile
SMILES ClC1=CC(OCC#N)=CC(Cl)=C1
Specifications
Melting Point 58°C to 60°C
Quantity 5 g
UN Number UN3439
Beilstein 4983677
Formula Weight 202.04
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorophenoxyacetonitrile