missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromoanisole, 97%, Thermo Scientific Chemicals

Product Code. 15432328
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432328 250 mg 250mg
15412328 1 g 1g
15422328 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15432328 Supplier Thermo Scientific Alfa Aesar Supplier No. H55925.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74137-36-3
Molecular Formula C7H6Br2O
Molecular Weight (g/mol) 265.932
MDL Number MFCD02258848
InChI Key OQZAQBGJENJMHT-UHFFFAOYSA-N
Synonym 3,5-dibromoanisole, 1,3-dibromo-5-methoxy-benzene, benzene, 1,3-dibromo-5-methoxy, pubchem2009, acmc-1bh5r, ksc493q1d, 1.3-dibromo-5-methoxybenzene
PubChem CID 11021812
IUPAC Name 1,3-dibromo-5-methoxybenzene
SMILES COC1=CC(=CC(=C1)Br)Br

Specifications

Melting Point 34°C to 38°C
Flash Point >110°C (230°F)
Quantity 250 mg
Formula Weight 265.93
Percent Purity 97%
Chemical Name or Material 3,5-Dibromoanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.