missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-2-hydroxypyridine, 97%, Thermo Scientific™

Product Code. p-7047866
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369865 1 g 1g
11379865 5 g 5g
11389865 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11369865 Supplier Thermo Scientific Alfa Aesar Supplier No. L19862.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-81-6
Molecular Formula C5H3Br2NO
Molecular Weight (g/mol) 252.893
MDL Number MFCD00023472
InChI Key XIFRODWVHSZAMM-UHFFFAOYSA-N
Synonym 3,5-dibromo-2-hydroxypyridine, 3,5-dibromopyridin-2-ol, 3,5-dibromo-2-pyridinol, 3,5-dibromo-2 1h-pyridinone, 2-hydroxy-3,5-dibromopyridine, 3,5-dibromo-2-pyridone, 3,5-dibromopyridine-2-ol, 2 1h-pyridinone, 3,5-dibromo, pubchem6448, acmc-209bwt
PubChem CID 642826
IUPAC Name 3,5-dibromo-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1Br)Br

Specifications

Melting Point ∼210°C
Odor Odorless
Quantity 1 g
Beilstein 119389
Formula Weight 252.89
Percent Purity 97%
Chemical Name or Material 3,5-Dibromo-2-hydroxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.