Learn More
3',5'-Di-O-acetyl-2'-deoxy-2'-fluorouridine, TRC
Specifications
Specifications
| Chemical Name or Material | 3',5'-Di-O-acetyl-2'-deoxy-2'-fluorouridine |
| CAS | 10212-13-2 |
| Molecular Formula | C13H15FN2O7 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C13H15FN2O7/c1-6(17)21-5-8-11(22-7(2)18)10(14)12(23-8)16-4-3-9(19)15-13(16)20/h3-4,8,10-12H,5H2,1-2H3,(H,15,19,20)/t8-,10-,11-,12-/m1/s1 |
| SMILES | CC(=O)OC[C@H]1O[C@H]([C@H](F)[C@@H]1OC(=O)C)N2C=CC(=O)NC2=O |
| IUPAC Name | [(2R,3R,4R,5R)-3-acetyloxy-5-(2,4-dioxopyrimidin-1-yl)-4-fluorooxolan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 330.27 |
| Formula Weight | 330.0863 |