Learn More
3,5-Bis(trifluoromethyl)-o-phenylenediamine, 97%
Product Code. 15439079
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15439079 1 g 1g
15449079 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15439079 Supplier Thermo Scientific Alfa Aesar Supplier No. L18180.03
 Restricted Item
In Stock
Add to basket
CAS: 367-65-7 | C8H6F6N2 | 244.14 g/mol
It is applied as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied as an active pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 367-65-7
Molecular Formula C8H6F6N2
Molecular Weight (g/mol) 244.14
MDL Number MFCD01631430
InChI Key BRLIJPMFMGTIAW-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl-1,2-diaminobenzene, 3,5-bis trifluoromethyl benzene-1,2-diamine, 3,5-bis trifluoromethyl-1,2-phenylenediamine, 1,2-diamino-3,5-bis trifluoromethyl benzene, 1,2-benzenediamine, 3,5-bis trifluoromethyl, acmc-1cr3d, 3,5-bis trifluoromethyl-o-phenylenediamine, 1,2-benzenediamine,3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenylene-1,2-diamine
PubChem CID 2773214
IUPAC Name 3,5-bis(trifluoromethyl)benzene-1,2-diamine
SMILES NC1=CC(=CC(=C1N)C(F)(F)F)C(F)(F)F
Specifications
Melting Point 44°C to 49°C
Quantity 1 g
UN Number UN2811
Beilstein 3344332
Solubility Information Insoluble in water.
Formula Weight 244.14
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)-o-phenylenediamine