Learn More
3,5-Bis(trifluoromethyl)-1H-1,2,4-triazole, TRC
Specifications
Specifications
| Chemical Name or Material | 3,5-Bis(trifluoromethyl)-1H-1,2,4-triazole |
| CAS | 709-62-6 |
| Molecular Formula | C4 H F6 N3 |
| Quantity | 100 mg |
| Synonym | 3,5-Bis(trifluoromethyl)-1,2,4-triazole, 3,5-Bis(trifluoromethyl)-s-triazole |
| InChI Formula | InChI=1S/C4HF6N3/c5-3(6,7)1-11-2(13-12-1)4(8,9)10/h(H,11,12,13) |
| SMILES | FC(F)(F)c1nnc([nH]1)C(F)(F)F |
| IUPAC Name | 3,5-bis(trifluoromethyl)-4H-1,2,4-triazole |
| Molecular Weight (g/mol) | 205.061 |
| Formula Weight | 205.007 |