Learn More
3,5,6-Trichlorosalicylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3,5,6-Trichloro-2-Hydroxybenzoic Acid |
| CAS | 40932-60-3 |
| Molecular Formula | C7 H3 Cl3 O3 |
| Quantity | 10 g |
| Synonym | 2,3,5-Trichloro-6-hydroxybenzoic acid, 3,5,6-Trichloro-2-hydroxybenzoic acid, 3,5,6-Trichlorosalicylic acid |
| InChI Formula | InChI=1S/C7H3Cl3O3/c8-2-1-3(9)6(11)4(5(2)10)7(12)13/h1,11H,(H,12,13) |
| SMILES | OC(=O)c1c(O)c(Cl)cc(Cl)c1Cl |
| IUPAC Name | 2,3,5-trichloro-6-hydroxybenzoic acid |
| Molecular Weight (g/mol) | 241.46 |
| Formula Weight | 239.9148 |