missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Morpholinyl)propyl-1-boronic acid pinacol ester, 98%, Thermo Scientific™

Product Code. 15437755
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437755 5 g 5g
15427755 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437755 Supplier Alfa Aesar™ Supplier No. H30191.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315281-09-4
Molecular Formula C13H26BNO3
Molecular Weight (g/mol) 255.165
MDL Number MFCD12546182
InChI Key CBGALTGWZUGSAO-UHFFFAOYSA-N
Synonym 3-4-morpholinyl propyl-1-boronic acid pinacol ester, 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl propyl morpholine
PubChem CID 73995111
IUPAC Name 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)propyl]morpholine
SMILES B1(OC(C(O1)(C)C)(C)C)CCCN2CCOCC2

Specifications

Refractive Index 1.4585
Quantity 5 g
Formula Weight 255.17
Percent Purity 98%
Chemical Name or Material 3-(4-Morpholinyl)propyl-1-boronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.