missing translation for 'onlineSavingsMsg'
Learn More

3,4-Methylenedioxybenzylamine, MP Biomedicals

Product Code. 11457995 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457995 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11457995 Supplier MP Biomedicals Supplier No. 0521697210

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2620-50-0
Molecular Formula C8H9NO2
MDL Number MFCD00005840
Quantity 10 g
Synonym piperonylamine, benzo d 1,3 dioxol-5-ylmethanamine, 3,4-methylenedioxybenzylamine, 1,3-benzodioxol-5-ylmethylamine, 1,3-benzodioxole-5-methanamine, 2h-1,3-benzodioxol-5-ylmethanamine, 3,4-methylenedioxy benzylamine, benzo-1,3-dioxole-5-methylamine, 1-1,3-benzodioxol-5-yl methanamine, 5-aminomethyl-1,3-benzodioxole
InChI Key ZILSBZLQGRBMOR-UHFFFAOYSA-N
SMILES NCC1=CC=C2OCOC2=C1
IUPAC Name 1-(2H-1,3-benzodioxol-5-yl)methanamine
Molecular Weight (g/mol) 151.17
PubChem CID 75799
Formula Weight 151.17
Chemical Name or Material 3, 4-Methylenedioxybenzonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.