Learn More
3,4-Methylenedioxy-alpha-Pyrrolidinohexanophenone Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 3,4-Methylenedioxy-alpha-Pyrrolidinohexanophenone (hydrochloride) |
| CAS | 24622-61-5 |
| Molecular Formula | C17 H23 N O3 . H Cl |
| Quantity | 2.5 mg |
| Synonym | 1-Hexanone, 1-(1,3-benzodioxol-5-yl)-2-(1-pyrrolidinyl)-, hydrochloride (1:1), 1-Hexanone, 1-(1,3-benzodioxol-5-yl)-2-(1-pyrrolidinyl)-, hydrochloride (9CI), Hexanophenone, 3',4'-(methylenedioxy)-2-(1-pyrrolidinyl)-, hydrochloride (8CI), 1-(3',4'-Methylenedioxyphenyl)-2-pyrrolidino-1-hexanone hydrochloride, 3,4-Methylenedioxy-α-pyrrolidinohexanophenone hydrochloride, 3,4-MD-α-PHP hydrochloride, 3,4-MDPHP hydrochloride |
| InChI Formula | InChI=1S/C17H23NO3.ClH/c1-2-3-6-14(18-9-4-5-10-18)17(19)13-7-8-15-16(11-13)21-12-20-15;/h7-8,11,14H,2-6,9-10,12H2,1H3;1H |
| SMILES | Cl.CCCCC(N1CCCC1)C(=O)c2ccc3OCOc3c2 |
| IUPAC Name | 1-(1,3-benzodioxol-5-yl)-2-pyrrolidin-1-ylhexan-1-one;hydrochloride |
| Molecular Weight (g/mol) | 325.83 |
| Formula Weight | 325.144 |