Learn More
3',4'-(Methylenedioxy)acetophenone, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 3162-29-6 |
| Molecular Formula | C9H8O3 |
| MDL Number | MFCD00005831 |
| Quantity | 500 g |
| SMILES | C1=C(C=CC2=C1OCO2)C(=O)C |
| Molecular Weight (g/mol) | 164.16 |
| Percent Purity | 98% |
| Chemical Name or Material | 3',4'-(Methylenedioxy)acetophenone |