missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluorophenyl)-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 15497287
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 15497287

Brand: Thermo Scientific Alfa Aesar H34355.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-(4-Fluorophenyl)-1H-pyrazole is used as pharmaceutical intermediates.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154258-82-9
Molecular Formula C9H7FN2
Molecular Weight (g/mol) 162.167
MDL Number MFCD00665857
InChI Key STTNBHIFTZEPSH-UHFFFAOYSA-N
Synonym 3-4-fluorophenyl-1h-pyrazole, 3-4-fluorophenyl pyrazole, 5-4-fluorophenyl-1h-pyrazole, 3-4-fluoro-phenyl-1h-pyrazole, 5-4-fluorophenyl pyrazole, 1h-pyrazole, 3-4-fluorophenyl, 3-4-fluorophenyl-2h-pyrazole, pubchem10424, acmc-1c1tf, acmc-2099xn
PubChem CID 688691
IUPAC Name 5-(4-fluorophenyl)-1H-pyrazole
SMILES C1=CC(=CC=C1C2=CC=NN2)F

Specifications

Melting Point 101°C to 103°C
Quantity 10 g
Formula Weight 162.17
Percent Purity 97%
Chemical Name or Material 3-(4-Fluorophenyl)-1H-pyrazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.