accessibility menu, dialog, popup

UserName

CAS RN 154258-82-9

NHNF

CAS RN 154258-82-9

IUPAC Name: 5-(4-fluorophenyl)-1H-pyrazole
Synonyms: 3-(4-fluorophenyl)-2H-pyrazole
Molecular Weight (g/mol): 162.17
Molecular Formula: C9H7FN2
InChi Key: STTNBHIFTZEPSH-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=CC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results