missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzaldehyde, TRC

Product Code. 30383933
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383933 5 g 5g
30371176 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383933 Supplier Toronto Research Chemicals Supplier No. TRCD4645355G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5973-71-7
Molecular Formula C9 H10 O
Molecular Weight (g/mol) 134.18
Synonym Benzaldehyde, 3,4-dimethyl-, 3,4-Dimethylbenzaldehyde (ACI), 3,4-DBAL
IUPAC Name 3,4-dimethylbenzaldehyde
SMILES Cc1ccc(C=O)cc1C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H10O/c1-7-3-4-9(6-10)5-8(7)2/h3-6H,1-2H3
Formula Weight 134.0732
Chemical Name or Material 3,4-Dimethylbenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.