Learn More
3,4-Dimethoxyphenethyl isothiocyanate, 95%, Thermo Scientific™
Product Code. 11370857
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11370857 10 g 10g
10007982 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11370857 Supplier Thermo Scientific Maybridge Supplier No. TL00669EA
 Restricted Item
Estimated Shipment: 08-12-2026
Chemical Identifiers
CAS 21714-25-0
Molecular Formula C11H13NO2S
Molecular Weight (g/mol) 223.29
MDL Number MFCD00041366
InChI Key GHYFLDWHIVKQLS-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenethyl isothiocyanate, 4-2-isothiocyanatoethyl-1,2-dimethoxybenzene, 2-3,4-dimethoxyphenyl ethyl isothiocyanate, benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy, 2-3,4-dimethoxyphenyl ethanisothiocyanate, acmc-20anep, 3,4-dimethoxyphenethylisothiocyanate, b-3,4-dimethoxyphenyl ethyl isothiocyanate, 3,4-dimethoxy-1-2-isothiocyanatoethyl benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy-benzene
PubChem CID 140856
IUPAC Name 4-(2-isothiocyanatoethyl)-1,2-dimethoxybenzene
SMILES COC1=CC=C(CCN=C=S)C=C1OC
Specifications
Assay Percent Range 95%
Quantity 10 g
Formula Weight 223.29
Chemical Name or Material 3, 4-Dimethoxyphenethyl isothiocyanate