missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzoic Acid, MP Biomedicals

Product Code. 11493245 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493245 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11493245 Supplier MP Biomedicals Supplier No. 0521403580

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 93-07-2
Molecular Formula C9H10O4
Quantity 100 g
Synonym veratric acid, benzoic acid, 3,4-dimethoxy, dimethylprotocatechuic acid, veratrylic acid, 3,4-dimethylprotocatechuic acid, veratrumenoic acid, 3,4-dimethoxy-benzoic acid, unii-1yy04e7rr4, 3,4-dimethoxybenzoes
InChI Key DAUAQNGYDSHRET-UHFFFAOYSA-N
SMILES COC1=C(C=C(C=C1)C(=O)O)OC
IUPAC Name 3,4-dimethoxybenzoic acid
Molecular Weight (g/mol) 182.175
PubChem CID 7121
ChEBI CHEBI:296881
Formula Weight 182.18
Chemical Name or Material 3, 4-Dimethoxybenzoic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.