Learn More
3,4-Dimethoxy-Alpha-[3-(methylamino)propyl]-Alpha-(1-methylethyl)-benzeneacetonitrile Hydrochloride, TRC
Product Code. 30370519
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30370519 10 mg 10mg
30385789 50 mg 50mg
30384107 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30370519 Supplier Toronto Research Chemicals Supplier No. TRCD46127510MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 67775-97-7
Molecular Formula C17 H26 N2 O2 . Cl H
Molecular Weight (g/mol) 326.86
Synonym (2RS)-2-(3,4-Dimethoxyphenyl)-5-(methylamino)-2-(1-methylethyl)pentanenitrile Hydrochloride, Verapamil Hydrochloride Imp. F (EP) as Hydrochloride, Benzeneacetonitrile, 3,4-dimethoxy-α-[3-(methylamino)propyl]-α-(1-methylethyl)-, hydrochloride (1:1) (ACI), Benzeneacetonitrile, 3,4-dimethoxy-α-[3-(methylamino)propyl]-α-(1-methylethyl)-, monohydrochloride (9CI), 3,4-Dimethoxy-α-[3-(methylamino)propyl]-α-(1-methylethyl)benzeneacetonitrile monohydrochloride
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-(methylamino)-2-propan-2-ylpentanenitrile;hydrochloride
SMILES Cl.CNCCCC(C#N)(C(C)C)c1ccc(OC)c(OC)c1
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C17H26N2O2.ClH/c1-13(2)17(12-18,9-6-10-19-3)14-7-8-15(20-4)16(11-14)21-5;/h7-8,11,13,19H,6,9-10H2,1-5H3;1H
Formula Weight 326.1761
Chemical Name or Material (2RS)-2-(3,4-Dimethoxyphenyl)-5-(methylamino)-2-(1-methylethyl)pentanenitrile Hydrochloride